Abnova
Manufacturer Code:H00055763A01
Catalog # 89-107-067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 780 a.a. to 879 a.a) partial recombinant protein with GST tag.,EXOC1 (NP_839955 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant EXOC1.
For Research Use Only
Protein Aliases: BM-102; Exocyst complex component 1; Exocyst complex component Sec3; FLJ10893; SEC3; SEC3-like 1; SEC3L1; SEC3P
Gene Aliases: BM-012; BM-102; EXOC1; SEC3; SEC3L1; SEC3P
UniProt ID: (Human) Q9NV70
Entrez Gene ID: (Human) 55763
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.