Invitrogen
Manufacturer Code:PA523980
Catalog # PIPA523980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 166-193 amino acids from the central region of human DEM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf176 Exo V defects in morphology protein 1 homolog probable exonuclease V; defects in morphology 1 homolog; Defects in morphology protein 1 homolog; Defects in morphology protein 1-like; Exo V; exonuclease 5; Exonuclease V; probable exonuclease V
Gene Aliases: 3110037I16Rik; AV297100; C1orf176; DEM1; Exo V; EXO5; hExo5; mExo5
UniProt ID: (Human) Q8WTW9, (Mouse) Q9CXP9
Entrez Gene ID: (Human) 64789, (Mouse) 73172, (Chinese hamster) 100753327
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.