Abnova
Manufacturer Code:H00009156K
Catalog # 89-202-996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human EXO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human EXO1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Exonuclease 1; Exonuclease I; HEX1 hExoI; hExo1; hExoI; rad2 nuclease family member, homolog of S. cerevisiae exonuclease 1
Gene Aliases: EXO1; EXOI; HEX1; hExoI
UniProt ID: (Human) Q9UQ84
Entrez Gene ID: (Human) 9156
Molecular Function:
DNA binding protein
damaged DNA-binding protein
endodeoxyribonuclease
exodeoxyribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.