Invitrogen
Manufacturer Code:PA5113397
Catalog # PIPA5113397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Exonuclease 3&prime-5&prime domain-containing protein 2 protein (137-245AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3&prime-5&prime exoribonuclease EXD2 4930539P14Rik C14orf114 C85658 Exd2 EXDL2 exonuclease 3&prime-5&prime domain containing 2 Exonuclease 3&prime-5&prime domain-containing protein 2 exonuclease 3&prime&apos-5&prime&apos domain-like 2 exonuclease 3&prime-5&prime domain-like 2 Exonuclease 3&prime-5&prime domain-like-containing protein 2 RGD1311087; 3'-5' exoribonuclease EXD2; exonuclease 3''-5'' domain-like 2; Exonuclease 3'-5' domain-containing protein 2; exonuclease 3'-5' domain-like 2; Exonuclease 3'-5' domain-like-containing protein 2
Gene Aliases: 4930539P14Rik; C14orf114; C85658; EXD2; EXDL2
UniProt ID: (Human) Q9NVH0, (Mouse) Q8VEG4
Entrez Gene ID: (Human) 55218, (Mouse) 97827
Molecular Function:
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.