OriGene
Manufacturer Code:TA502102
Catalog # 50-167-3186
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G1 |
Immunogen | Full length human recombit protein of human EXD1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EXDL1; Exonuclease 3'-5' domain-containing protein 1; Exonuclease 3'-5' domain-like-containing protein 1; Inactive exonuclease EXD1; piRNA biogenesis protein EXD1
Gene Aliases: EXD1; EXDL1
UniProt ID: (Human) Q8NHP7
Entrez Gene ID: (Human) 161829
Molecular Function:
RNA binding protein
exoribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.