Novus Biologicals
Manufacturer Code:NBP30479120UL
Catalog # NB093415
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1784-2033 of human EVPL (NP_001979.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 210 kDa cornified envelope precursor protein; 210 KDa Cornified Envelope Precursor Protein 210 KDa Paraneoplastic Pemphigus Antigen envoplakin EVPK p210; 210 kDa paraneoplastic pemphigus antigen; Envoplakin; p210
Gene Aliases: EVPK; EVPL
UniProt ID: (Human) Q92817
Entrez Gene ID: (Human) 2125
Molecular Function: cytoskeletal protein intermediate filament intermediate filament binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.