Novus Biologicals
Manufacturer Code:NBP193723
Catalog # NBP193723
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GFLSILYSIGSKPLQIWDKKVRNGHIKRITDNDIQSLVLEIEGTNVSTTYITCPADPKKTLGIKLPFLVMIIKNLKKYFTFEVQVLDDKNVRRRFRASNYQSTTRVKPFICTMPMRLDDGWNQIQFNLL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basal body up-regulated protein 22; C16orf80 chromosome 16 open reading frame 80 fSAP23 GTL3; Cilia- and flagella-associated protein 20; flagellar associated protein 20 homolog; functional spliceosome-associated protein 23; gene trap locus 3; Transcription factor IIB; UPF0468 protein C16orf80
Gene Aliases: BUG22; C16orf80; CFAP20; EVORF; fSAP23; GTL3
UniProt ID: (Human) Q9Y6A4
Entrez Gene ID: (Human) 29105
Molecular Function: nuclease nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.