Invitrogen
Manufacturer Code:MA514892
Catalog # PIMA514892
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | S.446.4 |
Immunogen | Synthetic peptide corresponding to residues surrounding Glu56 of human Evi-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AML1-EVI-1 fusion protein; AML1-EVI-1 fusion protein MDS1 and EVI1 complex locus protein EVI1 MDS1 and EVI1 complex locus protein MDS1 OTTHUMP00000213163 OTTHUMP00000213164 OTTHUMP00000213165 OTTHUMP00000213187 OTTHUMP00000213192 ecotropic viral integration site 1 ecotropic virus integration site 1 protein homolog myelodysplasia syndrome 1 protein myelodysplasia syndrome-associated protein 1 oncogene EVI1 zinc finger protein Evi1; Ecotropic virus integration site 1 protein homolog; EVI-1; Histone-lysine N-methyltransferase MECOM; MDS1 and EVI1 complex locus protein; MDS1 and EVI1 complex locus protein MDS1; Myelodysplasia syndrome 1 protein; Myelodysplasia syndrome-associated protein 1; oncogene EVI1; PR domain 3; zinc finger protein Evi1
Gene Aliases: AML1-EVI-1; EVI1; MDS1; MDS1-EVI1; MECOM; PRDM3; RUSAT2
UniProt ID: (Human) Q03112
Entrez Gene ID: (Human) 2122
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.