Invitrogen
Manufacturer Code:MA511142
Catalog # PIMA511142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 47AT612.84.88 |
Immunogen | Purified recombinant GST fusion protein encoding the N-terminal part of human EVI1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AML1-EVI-1 fusion protein; ecotropic viral integration site 1; Ecotropic virus integration site 1 protein; Ecotropic virus integration site 1 protein homolog; EVI-1; Histone-lysine N-methyltransferase MECOM; MDS1 and EVI1 complex locus protein; MDS1 and EVI1 complex locus protein EVI1; MDS1 and EVI1 complex locus protein MDS1; MDS1-EVI1; myelodysplasia syndrome 1 homolog; Myelodysplasia syndrome 1 protein; Myelodysplasia syndrome 1 protein homolog; Myelodysplasia syndrome-associated protein 1; oncogene EVI1; PR domain 3; zinc finger protein Evi1
Gene Aliases: AML1-EVI-1; D630039M04Rik; Evi-1; EVI1; Jbo; Mds; MDS1; MDS1-EVI1; MECOM; PRDM3; RUSAT2; Znfpr1b1
UniProt ID: (Human) Q03112, (Mouse) P14404
Entrez Gene ID: (Human) 2122, (Mouse) 14013
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.