Invitrogen
Manufacturer Code:PA5115561
Catalog # PIPA5115561
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MDS1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AML1-EVI-1 AML1-EVI-1 fusion protein D630039M04Rik ecotropic viral integration site 1 ecotropic virus integration site 1 protein Ecotropic virus integration site 1 protein homolog EVI1 EVI-1 Histone-lysine N-methyltransferase MECOM Jbo Mds Mds1 MDS1 and EVI1 complex locus MDS1 and EVI1 complex locus protein MDS1 and EVI1 complex locus protein EVI1 MDS1 and EVI1 complex locus protein MDS1 Mds1-Evi1 Mecom MGC163392 MGC97004 myelodysplasia syndrome 1 homolog Myelodysplasia syndrome 1 protein Myelodysplasia syndrome 1 protein homolog myelodysplasia syndrome-associated protein 1 oncogene EVI1 OTTHUMP00000213163 OTTHUMP00000213164 OTTHUMP00000213165 OTTHUMP00000213187 OTTHUMP00000213192 PR domain 3 PRDM3 RUSAT2 zinc finger protein Evi1 Znfpr1b1; AML1-EVI-1 fusion protein; ecotropic viral integration site 1; Ecotropic virus integration site 1 protein; Ecotropic virus integration site 1 protein homolog; EVI-1; Histone-lysine N-methyltransferase MECOM; MDS1 and EVI1 complex locus protein; MDS1 and EVI1 complex locus protein EVI1; MDS1 and EVI1 complex locus protein MDS1; myelodysplasia syndrome 1 homolog; Myelodysplasia syndrome 1 protein; Myelodysplasia syndrome 1 protein homolog; Myelodysplasia syndrome-associated protein 1; oncogene EVI1; PR domain 3; zinc finger protein Evi1
Gene Aliases: AML1-EVI-1; D630039M04Rik; Evi-1; EVI1; Jbo; Mds; MDS1; MDS1-EVI1; MECOM; PRDM3; RUSAT2; Znfpr1b1
UniProt ID: (Human) Q03112, (Mouse) P14404
Entrez Gene ID: (Human) 2122, (Mouse) 14013, (Rat) 294924
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.