R&D Systems
Manufacturer Code:MAB75061SP
Catalog # MAB75061SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 702321 |
Immunogen | <EM>E. coli</EM>-derived recombinant human EVI-1 Gly241-Met430 Accession # Q03112 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AML1-EVI-1 AML1-EVI-1 fusion protein ecotropic viral integration site 1 Ecotropic virus integration site 1 protein homolog EVI1 EVI-1 EVI1MDS1-EVI1 MDS1 MDS1 and EVI1 complex locus MDS1 and EVI1 complex locus protein EVI1 MDS1 and EVI1 complex locus protein MDS1 MECOM MGC163392 MGC97004 myelodysplasia syndrome 1 Myelodysplasia syndrome 1 protein Myelodysplasia syndrome-associated protein 1 oncogene EVI1 PRDM3 zinc finger protein Evi1; AML1-EVI-1 fusion protein; Ecotropic virus integration site 1 protein homolog; EVI-1; Histone-lysine N-methyltransferase MECOM; MDS1 and EVI1 complex locus protein; MDS1 and EVI1 complex locus protein MDS1; Myelodysplasia syndrome 1 protein; Myelodysplasia syndrome-associated protein 1; oncogene EVI1; PR domain 3; zinc finger protein Evi1
Gene Aliases: AML1-EVI-1; EVI1; MDS1; MDS1-EVI1; MECOM; PRDM3; RUSAT2
UniProt ID: (Human) Q03112
Entrez Gene ID: (Human) 2122
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.