Novus Biologicals
Manufacturer Code:NBP176903
Catalog # NBP176903
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 14 amino acid synthetic peptide from near the center of human EVER2. The immunogen is located within amino acids 300 - 350 of EVER2. Amino Acid Squence: WATKYSQDNKEESL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epidermodysplasia verruciformis 2; epidermodysplasia verruciformis 2 Epidermodysplasia verruciformis protein 2 EV2 EVER2MGC102701 EVIN2FLJ43684 FLJ40668 MGC40121 transmembrane channel-like 8 transmembrane channel-like protein 8; Epidermodysplasia verruciformis protein 2; transmembrane channel-like 8; Transmembrane channel-like protein 8
Gene Aliases: EV2; EVER2; EVIN2; TMC8
UniProt ID: (Human) Q8IU68
Entrez Gene ID: (Human) 147138
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.