Novus Biologicals
Manufacturer Code:NBP271938UV
Catalog # NBP271938UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B2 |
Immunogen | Recombinant protein fragment corresponding to amino acids 90-341 of human ETV7 (NP_057219) produced in E.coli. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Ets transcription factor TEL-2b; ETS translocation variant 7; ETS translocation variant 7 ets variant 7 ets variant gene 7 (TEL2 oncogene) TEL-2 TEL2 oncogene TEL2ETS-related protein Tel2 TELBEts transcription factor TEL-2b Tel-related Ets factor transcription factor ets transcription factor ETV7 Transcription factor Tel-2 TREF; ets variant gene 7 (TEL2 oncogene); ETS-related protein Tel2; Tel-related Ets factor; TEL2 oncogene; Transcription factor ETV7; Transcription factor Tel-2
Gene Aliases: ETV7; TEL-2; TEL2; TELB; TREF
UniProt ID: (Human) Q9Y603
Entrez Gene ID: (Human) 51513
Molecular Function: nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.