Invitrogen
Manufacturer Code:PA576040
Catalog # PIPA576040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human TEL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ETS translocation variant 6; ets variant gene 6 (TEL oncogene); ETS-related protein Tel1; Tel; TEL1 oncogene; Transcription factor ETV6; Transcription factor ETV6 ETS translocation variant 6 ETS-related protein Tel1 Tel ETV6 TEL TEL1
Gene Aliases: AW123102; AW557856; ETV6; TEL; TEL/ABL; TEL1; THC5
UniProt ID: (Human) P41212, (Mouse) P97360
Entrez Gene ID: (Human) 2120, (Mouse) 14011, (Rat) 312777
Molecular Function: nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.