Novus Biologicals
Manufacturer Code:NBP255689
Catalog # NBP255689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AAGPVQGVGPAPAPHSLPEPGPQQQTFAVPRPPHQPLQMPKMMPENQYPSEQRFQRQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ERMEts-related protein ERM ETS translocation variant 5 ets variant 5 ets variant gene 5 (ets-related molecule) ets-related molecule; ETS translocation variant 5; ets-related molecule; Ets-related protein ERM
Gene Aliases: ERM; ETV5
UniProt ID: (Human) P41161
Entrez Gene ID: (Human) 2119
Molecular Function: nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.