Invitrogen
Manufacturer Code:MA515646
Catalog # PIMA515646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3H3 |
Immunogen | Purified recombit fragment of human ETV5 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ERM ETV5; ETS translocation variant 5; ets variant gene 5; ets-related molecule; Ets-related protein ERM
Gene Aliases: 1110005E01Rik; 8430401F14Rik; ERM; ETV5
UniProt ID: (Human) P41161, (Mouse) Q9CXC9
Entrez Gene ID: (Human) 2119, (Mouse) 104156
Molecular Function:
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.