Invitrogen
Manufacturer Code:PA5100144
Catalog # PIPA5100144
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ETV5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110005E01Rik 8430401F14Rik ERM ETS translocation variant 5 ETS variant 5 ets variant gene 5 ets variant gene 5 (ets-related molecule) ets-related molecule ets-related protein ERM ETV5; ETS translocation variant 5; ets variant gene 5; ets variant gene 5 (ets-related molecule); ets-related molecule; Ets-related protein ERM
Gene Aliases: 1110005E01Rik; 8430401F14Rik; ERM; ETV5
UniProt ID: (Human) P41161, (Mouse) Q9CXC9
Entrez Gene ID: (Human) 2119, (Rat) 303828, (Mouse) 104156
Molecular Function:
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.