R&D Systems
Manufacturer Code:MAB9389SP
Catalog # MAB9389SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 815207 |
Immunogen | <em>E. coli</em>-derived recombinant human ETV1 Asn120-Leu190 Accession # P50549 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp781L0674 ER81 ER81ets variant gene 1 ETS translocation variant 1 ets variant 1 Ets-related protein 81 MGC104699 MGC120533 MGC120534; ETS translocation variant 1; ets variant gene 1; Ets-related protein 81
Gene Aliases: ER81; ETV1
UniProt ID: (Human) P50549
Entrez Gene ID: (Human) 2115
Molecular Function:
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.