Novus Biologicals
Manufacturer Code:NBP128868
Catalog # NBP128868
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C8B6 |
Immunogen | Purified recombinant fragment of ETV1(aa1-191) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp781L0674 ER81ets variant gene 1 ETS translocation variant 1 ets variant 1 Ets-related protein 81 MGC104699 MGC120533 MGC120534; ETS translocation variant 1; ets variant gene 1; Ets-related protein 81
Gene Aliases: ER81; ETV1
UniProt ID: (Human) P50549
Entrez Gene ID: (Human) 2115
Molecular Function:
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.