Invitrogen
Manufacturer Code:PA541484
Catalog # PIPA541484
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ETV1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: er81; ets related protein 81; ETS transcription factor Er81; ETS translocation variant 1; ETS variant 1; Ets variant 1 DKFZp781L0674 ER81 MGC104699 MGC120533 MGC120534; ets variant gene 1; Ets-related protein 81
Gene Aliases: ER81; Etsrp81; ETV1
UniProt ID: (Bovine) Q2KIC2, (Human) P50549, (Mouse) P41164
Entrez Gene ID: (Dog) 475246, (Bovine) 540846, (Human) 2115, (Horse) 100065470, (Rabbit) 100355978, (Rat) 362733, (Guinea pig) 100721423, (Mouse) 14009, (Zebrafish) 563876
Molecular Function:
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.