Novus Biologicals
Manufacturer Code:NBP146178
Catalog # NBP146178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 50 and 100 of human Tyrosyl DNA Phosphodiesterase-2 using the numbering given in entry NP_057698.2 (GeneID 51567). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-Tyr-DNA phosphodiesterase; 5'-tyrosyl-DNA phosphodiesterase; AD022 dJ30M3.3 EAP2 EAPII EC 3.1.4.- ETS1-associated protein 2 ETS1-associated protein II MGC1110215'-Tyr-DNA phosphodiesterase MGC90995'-tyrosyl-DNA phosphodiesterase TRAF and TNF receptor associated protein TTRAPTRAF and TNF receptor-associated protein Tyr-DNA phosphodiesterase 2 tyrosyl-DNA phosphodiesterase 2; EAPII; ETS1-associated protein 2; ETS1-associated protein II; TRAF and TNF receptor associated protein; TRAF and TNF receptor-associated protein; Tyr-DNA phosphodiesterase 2; Tyrosyl-DNA phosphodiesterase 2; Tyrosyl-RNA phosphodiesterase; VPg unlinkase
Gene Aliases: AD-022; AD022; dJ30M3.3; EAP2; EAPII; hTDP2; TDP2; TTRAP
UniProt ID: (Human) O95551
Entrez Gene ID: (Human) 51567
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.