Invitrogen
Manufacturer Code:PA5113882
Catalog # PIPA5113882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human AGXT2L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1300019H02Rik AGXT2L1 AI195447 Alanine--glyoxylate aminotransferase 2-like 1 alanine-glyoxylate aminotransferase 2-like 1 ethanolamine phosphate phospholyase Ethanolamine-phosphate phospho-lyase Etnppl; Alanine--glyoxylate aminotransferase 2-like 1; alanine-glyoxylate aminotransferase 2-like 1; Ethanolamine-phosphate phospho-lyase
Gene Aliases: AGXT2L1; ETNPPL
UniProt ID: (Human) Q8TBG4
Entrez Gene ID: (Human) 64850
Molecular Function: transaminase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.