Invitrogen
Manufacturer Code:PA538807
Catalog # PIPA538807
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human EKI2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EKI 2; EKI2 HMFT1716 EKI 2 ethanolamine kinase-like protein; Ethanolamine kinase 2; Ethanolamine kinase-like protein; putative ethanolamine kinase
Gene Aliases: 4933417N20Rik; AI197444; EKI 2; EKI2; ETNK2; HMFT1716; Tuc1
UniProt ID: (Human) Q9NVF9, (Mouse) A7MCT6
Entrez Gene ID: (Human) 55224, (Mouse) 214253
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.