Invitrogen
Manufacturer Code:PA513823
Catalog # PIPA513823
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 356-386 amino acids from the C-terminal region of human Ethanolamine Kinase (EKI1) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EKI 1; Ethanolamine kinase 1; Ethanolamine kinase EKI ETNK1; putative protein product of Nbla10396
Gene Aliases: 1110061E11Rik; 4930555L11Rik; AI195356; AI841245; C80956; D6Ertd3e; EKI; EKI 1; EKI1; ETNK1; I79_004753; Nbla10396
UniProt ID: (Human) Q9HBU6, (Mouse) Q9D4V0
Entrez Gene ID: (Human) 55500, (Chinese hamster) 100765795, (Mouse) 75320
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.