Invitrogen
Manufacturer Code:PA5109696
Catalog # PIPA5109696
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 100-300 of human ETNK1 (NP_061108.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110061E11Rik 4930555L11Rik AI195356 AI841245 C80956 D6Ertd3e EKI EKI 1 Eki1 Ethanolamine kinase Ethanolamine kinase 1 ETNK1 Nbla10396; EKI 1; Ethanolamine kinase 1; putative protein product of Nbla10396
Gene Aliases: EKI; EKI 1; EKI1; ETNK1; Nbla10396
UniProt ID: (Human) Q9HBU6
Entrez Gene ID: (Human) 55500
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.