Invitrogen
Manufacturer Code:MA515634
Catalog # PIMA515634
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C6 |
Immunogen | Purified recombit fragment of human BMX expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Bone marrow tyrosine kinase gene in chromosome X protein; BTK-like on X chromosome; Cytoplasmic tyrosine-protein kinase BMX; Epithelial and endothelial tyrosine kinase; ETK; ETK PSCTK2 PSCTK3 BMX; Etk/Bmx cytosolic tyrosine kinase; NTK38; NTK38 tyrosine kinase
Gene Aliases: BMX; ETK; PSCTK2; PSCTK3
UniProt ID: (Human) P51813
Entrez Gene ID: (Human) 660
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.