Invitrogen
Manufacturer Code:PA599532
Catalog # PIPA599532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ETK corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bmx BMX non-receptor tyrosine kinase bone marrow tyrosine kinase gene in chromosome X protein Bone marrow tyrosine kinase gene in chromosome X protein homolog BTK-like on X chromosome Cytoplasmic tyrosine-protein kinase BMX epa3 Epithelial and endothelial tyrosine kinase ETK Etk/Bmx Etk/Bmx cytosolic tyrosine kinase NTK38 NTK38 tyrosine kinase PSCTK2 PSCTK3 RGD1565643 Tyro8 TYRO8 protein tyrosine kinase 8; Bone marrow tyrosine kinase gene in chromosome X protein; Bone marrow tyrosine kinase gene in chromosome X protein homolog; BTK-like on X chromosome; Cytoplasmic tyrosine-protein kinase BMX; Epithelial and endothelial tyrosine kinase; ETK; Etk/Bmx cytosolic tyrosine kinase; NTK38; NTK38 tyrosine kinase; TYRO8 protein tyrosine kinase 8
Gene Aliases: BMX; ETK; Etk/Bmx; PSCTK2; PSCTK3; Tyro8
UniProt ID: (Human) P51813, (Mouse) P97504
Entrez Gene ID: (Human) 660, (Mouse) 12169
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.