Invitrogen
Manufacturer Code:PA514769
Catalog # PIPA514769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 212-242 amino acids from the central region of human BMX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bone marrow tyrosine kinase gene in chromosome X protein; Bone marrow tyrosine kinase gene in chromosome X protein homolog; BTK-like on X chromosome; Cytoplasmic tyrosine-protein kinase BMX; Cytoplasmic tyrosine-protein kinase BMX Bone marrow kinase BMX Epithelial and endothelial tyrosine kinase ETK NTK38; Epithelial and endothelial tyrosine kinase; ETK; Etk/Bmx cytosolic tyrosine kinase; NTK38; NTK38 tyrosine kinase; TYRO8 protein tyrosine kinase 8
Gene Aliases: BMX; ETK; Etk/Bmx; PSCTK2; PSCTK3; Tyro8
UniProt ID: (Human) P51813, (Mouse) P97504
Entrez Gene ID: (Human) 660, (Mouse) 12169
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.