Novus Biologicals
Manufacturer Code:NBP29030050UL
Catalog # NB022064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | Recombinant Human ETHE1 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: D83198 EC 3.1.2.6 ethylmalonic encephalopathy 1 Ethylmalonic encephalopathy protein 1 Hepatoma subtracted clone one protein HSCOEC 3.- protein ETHE1 mitochondrial YF13H12; ethylmalonic encephalopathy 1; Ethylmalonic encephalopathy protein 1; Hepatoma subtracted clone one protein; Persulfide dioxygenase ETHE1, mitochondrial; protein ETHE1, mitochondrial; Sulfur dioxygenase ETHE1
Gene Aliases: ETHE1; HSCO; YF13H12
UniProt ID: (Human) O95571
Entrez Gene ID: (Human) 23474
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.