Invitrogen
Manufacturer Code:PA528938
Catalog # PIPA528938
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 254 of Human ETHE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ethylmalonic encephalopathy 1; Ethylmalonic encephalopathy protein 1; Hepatoma subtracted clone one protein; HSCO YF13H12 ethylmalonic encephalopathy protein 1 hepatoma subtracted clone one protein protein ETHE1 mitochondrial; Persulfide dioxygenase ETHE1, mitochondrial; protein ETHE1, mitochondrial; Sulfur dioxygenase ETHE1
Gene Aliases: ETHE1; HSCO; YF13H12
UniProt ID: (Human) O95571
Entrez Gene ID: (Human) 23474
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.