Invitrogen
Manufacturer Code:PA513593
Catalog # PIPA513593
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 180-209 amino acids from the C-terminal region of human ETHE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ethylmalonic encephalopathy 1; Ethylmalonic encephalopathy protein 1; Ethylmalonic encephalopathy protein 1 homolog; Hepatoma subtracted clone one protein; Persulfide dioxygenase ETHE1, mitochondrial; Protein ETHE1 mitochondrial Ethylmalonic encephalopathy protein 1 Hepatoma subtracted clone one protein HSCO ETHE1; protein ETHE1, mitochondrial; Sulfur dioxygenase ETHE1
Gene Aliases: 0610025L15Rik; ETHE1; HSCO; YF13H12
UniProt ID: (Human) O95571, (Mouse) Q9DCM0
Entrez Gene ID: (Human) 23474, (Mouse) 66071
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.