Invitrogen
Manufacturer Code:PA572484
Catalog # PIPA572484
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 32-61 amino acids from the N-terminal region of human ETFDH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Electron transfer flavoprotein-ubiquinone oxidoreductase mitochondrial ETF-QO ETF-ubiquinone oxidoreductase Electron-transferring-flavoprotein dehydrogenase ETF dehydrogenase ETFDH; Electron transfer flavoprotein-ubiquinone oxidoreductase, mitochondrial; Electron-transferring-flavoprotein dehydrogenase; ETF dehydrogenase; ETF-QO; ETF-ubiquinone oxidoreductase
Gene Aliases: ETFDH; ETFQO; MADD
UniProt ID: (Human) Q16134
Entrez Gene ID: (Human) 2110
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.