Invitrogen
Manufacturer Code:PA519340
Catalog # PIPA519340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (EHDQPAHLTLRD) corresponding to the internal amino acids of ETFDH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Electron transfer flavoprotein-ubiquinone oxidoreductase, mitochondrial; electron transferring flavoprotein dehydrogenase; Electron-transferring-flavoprotein dehydrogenase; ETF dehydrogenase; ETF-QO; ETF-ubiquinone oxidoreductase; ETFDH electron-transferring-flavoprotein dehydrogenase ETFQO MADD ETF dehydrogenase ETF-ubiquinone oxidoreductase electron transfer flavoprotein ubiquinone oxidoreductase
Gene Aliases: 0610010I20Rik; AV001013; ETFDH; ETFQO; MADD
UniProt ID: (Human) Q16134, (Mouse) Q921G7
Entrez Gene ID: (Human) 2110, (Rat) 295143, (Mouse) 66841
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.