Invitrogen
Manufacturer Code:PA519336
Catalog # PIPA519336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QVTLEGDKLKVERE) corresponding to the internal amino acids of ETFB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-ETF; electron transfer flavoprotein beta-subunit; Electron transfer flavoprotein subunit beta; electron transfer flavoprotein, beta polypeptide; electron transferring flavoprotein, beta polypeptide; electron-transfer-flavoprotein, beta polypeptide; electron-transferring-flavoprotein, beta polypeptide; ETFB electron-transfer-flavoprotein beta polypeptide FP585 MADD electron transfer flavoprotein beta subunit electron transfer flavoprotein beta-subunit electron transfer flavoprotein beta polypeptide electron-transferring-flavoprotein beta polypeptide
Gene Aliases: 0610009I16Rik; 2810441H06Rik; ETFB; FP585; MADD
UniProt ID: (Human) P38117, (Rat) Q68FU3, (Mouse) Q9DCW4
Entrez Gene ID: (Human) 2109, (Rat) 292845, (Mouse) 110826
Molecular Function:
hydroxylase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.