Novus Biologicals
Manufacturer Code:NBP297524FR
Catalog # NB097365
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human ETFB fragment. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-ETF; beta-ETF electron transfer flavoprotein beta subunit electron transfer flavoprotein beta-subunit electron transfer flavoprotein subunit beta electron transfer flavoprotein beta polypeptide electron-transfer-flavoprotein beta polypeptide electron-transferring-flavoprotein beta polypeptide MADD; electron transfer flavoprotein beta-subunit; Electron transfer flavoprotein subunit beta; electron transfer flavoprotein, beta polypeptide; electron-transfer-flavoprotein, beta polypeptide; electron-transferring-flavoprotein, beta polypeptide
Gene Aliases: ETFB; FP585; MADD
UniProt ID: (Human) P38117
Entrez Gene ID: (Human) 2109
Molecular Function:
hydroxylase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.