Invitrogen
Manufacturer Code:PA571474
Catalog # PIPA571474
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 276-304 amino acids from the C-terminal region of human ETFA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-ETF; electron transfer flavoprotein alpha-subunit; Electron transfer flavoprotein subunit alpha, mitochondrial; electron transfer flavoprotein, alpha polypeptide; electron-transfer-flavoprotein alpha polypeptide; EMA GA2 MADD alpha-ETF electron transfer flavoprotein alpha-subunit electron transfer flavoprotein subunit alpha mitochondrial electron transfer flavoprotein alpha polypeptide glutaric aciduria II; glutaric aciduria II
Gene Aliases: 2010200I21Rik; D9Ertd394e; EMA; ETFA; GA2; MADD
UniProt ID: (Human) P13804, (Mouse) Q99LC5
Entrez Gene ID: (Human) 2108, (Mouse) 110842
Molecular Function: dehydrogenase oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.