Novus Biologicals
Manufacturer Code:NBP152058
Catalog # NBP152058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KSPDTFVRTIYAGN corresponding to internal region according to NP_001121188.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-ETF; alpha-ETF electron transfer flavoprotein alpha-subunit Electron transfer flavoprotein subunit alpha mitochondrial electron-transfer-flavoprotein alpha polypeptide EMA GA2 glutaric aciduria II MADD; electron transfer flavoprotein alpha-subunit; Electron transfer flavoprotein subunit alpha, mitochondrial; electron transfer flavoprotein, alpha polypeptide; electron-transfer-flavoprotein alpha polypeptide; glutaric aciduria II
Gene Aliases: EMA; ETFA; GA2; MADD
UniProt ID: (Human) P13804
Entrez Gene ID: (Human) 2108
Molecular Function: dehydrogenase oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.