Invitrogen
Manufacturer Code:PA530463
Catalog # PIPA530463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 176 and 393 of Human ETEA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ETEA UBXD8 UBXN3B FAS-associated factor 2 UBX domain containing 8 UBX domain protein 3B UBX domain-containing protein 3B UBX domain-containing protein 8 expressed in T-cells and eosinophils in atopic dermatitis; expressed in T-cells and eosinophils in atopic dermatitis; FAS-associated factor 2; Protein ETEA; UBX domain containing 8; UBX domain protein 3B; UBX domain-containing protein 3B; UBX domain-containing protein 8
Gene Aliases: ETEA; FAF2; KIAA0887; UBXD8; UBXN3B
UniProt ID: (Human) Q96CS3
Entrez Gene ID: (Human) 23197
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.