Invitrogen
Manufacturer Code:PA537162
Catalog # PIPA537162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in human ETAA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ETAA16; Ewing tumor-associated antigen 1; Ewing&primes tumor-associated antigen 1 Ewing&primes tumor-associated antigen 16 ETAA16; Ewing's tumor-associated antigen 1; Ewing's tumor-associated antigen 16
Gene Aliases: ETAA1; ETAA16; RGD1560957
UniProt ID: (Human) Q9NY74
Entrez Gene ID: (Human) 54465, (Rat) 498420
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.