OriGene
Manufacturer Code:CF505036
Catalog # 50-166-3332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5G8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 24-263 of human ESRRG produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ERR gamma-2; ERR3 ERRgamma NR3B3; Estrogen receptor-related protein 3; Estrogen-related receptor gamma; Nuclear receptor subfamily 3 group B member 3
Gene Aliases: ERR3; ERRG2; ERRgamma; ESRRG; KIAA0832; NR3B3
UniProt ID: (Human) P62508
Entrez Gene ID: (Human) 2104
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.