Invitrogen
Manufacturer Code:PA533525
Catalog # PIPA533525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 14 amino acid peptide from N-terminus of human ESRRG / ERR3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ERR gamma-2; ESRRG ERR Gamma ERR gamma-2 ERR3 Err-gamma NR3B3 ERRG2 ERRgamma KIAA0832; Estrogen receptor-related protein 3; Estrogen-related receptor gamma; Nuclear receptor subfamily 3 group B member 3
Gene Aliases: ERR3; ERRG2; ERRgamma; ESRRG; KIAA0832; NR3B3
UniProt ID: (Human) P62508
Entrez Gene ID: (Human) 2104
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.