Invitrogen
Manufacturer Code:PA527755
Catalog # PIPA527755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment contain a sequence corresponding to a region within amino acids 53 and 435 of ERR gamma |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ERR gamma-2; ERR3 ERRgamma NR3B3 ERR gamma-2 estrogen receptor-related protein 3 nuclear receptor subfamily 3 group B member 3; Estrogen receptor-related protein 3; estrogen-related receptor 3; Estrogen-related receptor gamma; Nuclear receptor subfamily 3 group B member 3
Gene Aliases: ERR3; Errg; ERRG2; ERRgamma; ESRRG; KIAA0832; mKIAA0832; NR3B3
UniProt ID: (Human) P62508, (Mouse) P62509, (Rat) P62510
Entrez Gene ID: (Human) 2104, (Mouse) 26381, (Rat) 360896
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.