Proteintech
Manufacturer Code:226441AP150UL
Catalog # 50-173-3910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ESRRB Fusion Protein Ag18523 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DFNB35 ERR2 ERRb ERRbeta ERRbeta 2 ESRL2 ESRRB ESRRB protein estrogen related receptor beta NR3B2; ERR beta-2; ERR-beta; ERRbeta-2; estrogen receptor related 2; Estrogen receptor-like 2; estrogen-related nuclear receptor beta; Estrogen-related receptor beta; estrogen-related receptor, beta; nuclear receptor ERRB2; Nuclear receptor subfamily 3 group B member 2; orphan nuclear receptor; Steroid hormone receptor ERR2
Gene Aliases: DFNB35; Err-2; ERR2; ERRb; ERRB2; ESRL2; ESRRB; Estrrb; NR3B2
UniProt ID: (Human) O95718, (Mouse) Q61539, (Rat) P11475
Entrez Gene ID: (Human) 2103, (Mouse) 26380, (Rat) 299210
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.