OriGene
Manufacturer Code:TA808634
Catalog # 50-168-2267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9D10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-205 of human ESRRA produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ERR-alpha; ERR1 ERRa ERRalpha ESRL1 NR3B1; Estrogen receptor-like 1; estrogen-related nuclear receptor alpha; Estrogen-related receptor alpha; Nuclear receptor subfamily 3 group B member 1; Steroid hormone receptor ERR1
Gene Aliases: ERR1; ERRa; ERRalpha; ESRL1; ESRRA; NR3B1
UniProt ID: (Human) P11474
Entrez Gene ID: (Human) 2101
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.