Invitrogen
Manufacturer Code:PA5113717
Catalog # PIPA5113717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal region of human RBM35B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9530027K23Rik Epithelial splicing regulatory protein 2 Esrp2 PP7059 Rbm35b RNA binding motif protein 35A RNA binding motif protein 35b RNA-binding motif protein 35B RNA-binding protein 35B; Epithelial splicing regulatory protein 2; RNA binding motif protein 35A; RNA binding motif protein 35B; RNA-binding motif protein 35B; RNA-binding protein 35B
Gene Aliases: ESRP2; PP7059; RBM35B
UniProt ID: (Human) Q9H6T0
Entrez Gene ID: (Human) 80004
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.