OriGene
Manufacturer Code:TA801038
Catalog # 50-167-8663
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E12 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-260 of human ESR2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ER beta; ER-beta; ER-BETA ESR-BETA ESRB ESTRB Erb NR3A2; estrogen receptor 2 (ER beta); Estrogen receptor beta; estrogen receptor beta 4; Nuclear receptor subfamily 3 group A member 2
Gene Aliases: ER-BETA; Erb; ESR-BETA; ESR2; ESRB; ESTRB; NR3A2
UniProt ID: (Human) Q92731
Entrez Gene ID: (Human) 2100
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.