Proteintech
Manufacturer Code:140071AP150UL
Catalog # 50-172-9150
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ESR2 Fusion Protein Ag5103 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ER beta; ER BETA Erb ESR BETA ESR2 ESRB ESTRB estrogen receptor 2 (ER beta) Estrogen receptor beta NR3A2; ER-beta; estrogen receptor 2 (ER beta); estrogen receptor 2 beta; Estrogen receptor beta; estrogen receptor beta 4; Nuclear receptor subfamily 3 group A member 2; oestrogen receptor beta
Gene Aliases: ER-BETA; ER[b]; Erb; Erb2; ERbeta; ESR-BETA; ESR2; ESRB; ESTRB; NR3A2
UniProt ID: (Human) Q92731, (Mouse) O08537, (Rat) Q62986
Entrez Gene ID: (Human) 2100, (Mouse) 13983, (Rat) 25149
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.