Proteintech
Manufacturer Code:207171AP150UL
Catalog # 50-173-3050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actin cytoskeletal regulatory protein; Autosomal recessive deafness type 36 protein; DFNB36 espin ESPN; Ectoplasmic specialization protein; Espin; jerker; Jerker, deafness locus
Gene Aliases: DFNB36; ESPN; Je; LP2654
UniProt ID: (Human) B1AK53, (Rat) Q63618, (Mouse) Q9ET47
Entrez Gene ID: (Human) 83715, (Rat) 56227, (Mouse) 56226
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.