Proteintech
Manufacturer Code:234961AP150UL
Catalog # 50-173-4084
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ESF1 Fusion Protein Ag19037 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABT1 associated protein ABTAP C20orf6 ESF1 ESF1 homolog HDCMC28P; ABT1-associated protein; ESF1 homolog; ESF1, nucleolar pre-rRNA processing protein, homolog
Gene Aliases: 2610101J03Rik; ABTAP; AW545818; bA526K24.1; C20orf6; C79684; ESF1; HDCMC28P
UniProt ID: (Human) Q9H501, (Mouse) Q3V1V3
Entrez Gene ID: (Human) 51575, (Mouse) 66580
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.