Invitrogen
Manufacturer Code:PA530020
Catalog # PIPA530020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 42 and 258 of Human ESD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Esterase 10; Esterase D; esterase D/formylglutathione hydrolase; FGH; FGH S-formylglutathione hydrolase esterase 10 esterase D/formylglutathione hydrolase methylumbelliferyl-acetate deacetylase; Methylumbelliferyl-acetate deacetylase; S-formylglutathione hydrolase; Sid 478; testicular tissue protein Li 66
Gene Aliases: Es-10; Es10; ESD; FGH; Sid478
UniProt ID: (Human) P10768, (Mouse) Q9R0P3
Entrez Gene ID: (Human) 2098, (Mouse) 13885
Molecular Function: esterase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.